Drip pads
Supplier: JUSTRITE MANUFACTURING
28460
JUSU28460EA 68.5 SGD
JUSU28460
Drip pads
Spill Control Materials
Drip pads can capture small oil leaks and drips before they contaminate the ground or create unsafe work surfaces.
  • Lightweight, flexible, and portable
  • Made from UV and chemical-resistant modified PVC
  • Coated mesh outer layer to trap coarse debris
These drip pads feature replaceable, oil-only sorbent with a welded leakproof barrier. Rated for drive over applications, pads include brass grommets in opposing corners for anchoring. Anchoring stakes not included. Pads come in two sizes; extended model is weighted to keep it down in high winds. Outer shellMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable