Containment berms, QuickBerm® Lite
Supplier: JUSTRITE MANUFACTURING
28556
JUSU28556EA 1180 SGD
JUSU28556
Containment berms, QuickBerm® Lite
Spill Control Materials
Portable, versatile containment berm deploys quickly for emergency spill response.
  • Leakproof, tough Durable EnGuard™I, PVC coated fabric provides broad UV and chemical resistance that withstands repeated usage
  • Reinforced inside supports reduce sagging and prevent tripping hazards that can be caused by outside supports
Lightweight berm folds down easily to a small size for storage in trucks, emergency vehicles, and spill kits. Add optional grates to help reduce contact with contaminated liquids by personnel or equipment.
Certifications: Helps toMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable