Spill control pallets, EcoPolyBlend™
Supplier: JUSTRITE MANUFACTURING
28637 28635 28623 28625
JUSU28637EA 1010 SGD
JUSU28637 JUSU28635 JUSU28623 JUSU28625
Spill control pallets, EcoPolyBlend™
Drum Storage
These pallets are made of up to 100% recycled polyethylene; dirt hiding black colour is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack.
  • Independently validated by UL Environment
  • Seamless, robust design eliminates leaks
  • Excellent chemical compatibility and no corrosion
  • Sturdy and self-locating grates remove for easy cleaning
  • Square or space saving linear styles in multiple sizes offer storage
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable