Spill containment caddy, EcoPolyBlend™
Supplier: JUSTRITE MANUFACTURING
28665
JUSU28665EA 3300 SGD
JUSU28665
Spill containment caddy, EcoPolyBlend™
Drum Storage
Made up of 90% recycled polyethylene.
  • Single-piece, double-walled seamless design eliminates leaks
  • Ergonomic design with tilt/stop feature minimises back strain
  • Rubber, 254 mm wheels roll over bumpy terrain use indoors or out
They offer convenient handling and dispensing of hazardous chemicals with complete assurance of compliance with EPA and SPCC regulations. It is the ideal choice for managing either 110 or 200 litre drums and the nylon strap holds drum securely in place.