Spill control drum management system, EcoPolyBlend™
Supplier: JUSTRITE MANUFACTURING
28667 28669 28671
JUSU28667EA 2580 SGD
JUSU28667 JUSU28669 JUSU28671
Spill control drum management system, EcoPolyBlend™
Drum Storage
EcoPolyBlend™ drum management system is environmentally friendly-made of recycled, durable polyethylene stands up to the harshest environments.
  • Space saving and stackable
  • Chemical resistant
  • Molded-in drum lock-channels hold drums in place
It consists of three parts such as a base module, stack module and dispensing shelf.
Base module holds two drums with a 1388 kg total load capacity. Easily relocate the base module with a forklift when empty. The 250 l open-view containment sump makes it easy to spot any spills for a quick cleanup.