Spill control funnels, EcoPolyBlend™
Supplier: JUSTRITE MANUFACTURING
28680
JUSU28680EA 161 SGD
JUSU28680
Spill control funnels, EcoPolyBlend™
Funnels Filtration Funnels Drum Funnels
Spill control funnel is an eco-friendly, cost-effective way to collect liquids and keep drum tops clean.
  • Low-profile and wide diameter
  • Quick and easy pouring
  • Molded-in hook stores optional funnel cover
Scalloped, sloped design minimizes splashes and prevents containers from resting in their own liquids. Large, flat welll at the throat of funnel allows for passive draining of paint tins, filters, buckets and other containers.
OD×H: 533×83 mm; Capacity: 2 kg