Spill trays, EcoPolyBlend™
Supplier: JUSTRITE MANUFACTURING
28715
JUSU28715EA 174 SGD
JUSU28715
Spill trays, EcoPolyBlend™
Trays/Dishes/Pots Spillage Trays/Pans
Eco-friendly spill trays are made from 100% post-industrial recycled PE will not rust or corrode.
  • Lightweight, low profile design for small-scale spill control
  • Raised rib bottom adds rigidity and keeps contents out of spills
  • Rated for service temperatures between –34 to +49 °C
Perfect for pouring and filling applications. Ideal for containing leaks from small equipment or batteries.