Bottle Jacket
Supplier: JUSTRITE MANUFACTURING
56610-100
JUSU56610-100EA 0 SGD
JUSU56610-100
Bottle Jacket
Carriers Bottle Carriers/Containers
Bottle jacket gives the added protection of a polyethylene bottle without contaminating high purity solvents
Fits 4 L and 1 gal. bottles. Lab bottles fit to the bottom half of the cover, and lid twists securely for safe pouring. Jacket also acts as a shock absorber, minimizing breakage and spills due to bumps or falls.