Safety cans, type II AccuFlow™, galvanised steel
Supplier: JUSTRITE MANUFACTURING
7250120 7210120Z 7225130Z 7220120Z 7250130Z 7220220Z 7250230Z
JUSU7250120EA 260 SGD
JUSU7250120 JUSU7210120Z JUSU7225130Z JUSU7220120Z JUSU7250130Z JUSU7220220Z JUSU7250230Z
Safety cans, type II AccuFlow™, galvanised steel
Safety Cans
Safety cans are designed to convenient and safe storage and dispense of flammable liquids.
  • Rounded steel comfort handle protects hand injury
  • Trilingual warning label in English/Spanish/French
  • Large ID zone identifies contents to reduce misuse
Features vented AccuFlow™ manifold and Safe-Squeeze® trigger handle provides the controlled liquid flow ever and flexible metal hose of 229 mm long to reach small openings. Ergonomically designed quick-fill port with comfort lift lever makes filling easier than ever.
Fill port is equipped with a long length,More Product Information