Safety storage cabinets for flammables
Supplier: JUSTRITE MANUFACTURING
882404 882414 882424 882434 883004 883024 883604 883624 884804 884824 884807 882400 882407 882410 882417 882420 882427 882430 882437 883000 883007 883020 883027 883600 883607 883620 883627 884800 884820 884827
JUSU882404EA 4300 SGD
JUSU882404 JUSU882414 JUSU882424 JUSU882434 JUSU883004 JUSU883024 JUSU883604 JUSU883624 JUSU884804 JUSU884824 JUSU884807 JUSU882400 JUSU882407 JUSU882410 JUSU882417 JUSU882420 JUSU882427 JUSU882430 JUSU882437 JUSU883000 JUSU883007 JUSU883020 JUSU883027 JUSU883600 JUSU883607 JUSU883620 JUSU883627 JUSU884800 JUSU884820 JUSU884827
Safety storage cabinets for flammables
Furniture Storage Cabinets
Ideal in laboratories under fume hoods, benches, or counters for safe storage and venting of solvents and flammable liquids.
  • Stainless steel, 3 point bullet self-latching system
  • Leak proof bottom sump to capture spills
  • SpillSlope™ adjustable shelves
  • U-Loc™ handle comes with double key set or accepts optional padlock
  • Highly reflective Haz-Alert™ hazard labeling at top and bottom
Constructed of fully welded, double-wall 18 gauge steel with insulating air space. Finished in a durable, chemical resistant powder paint, these cabinets come in three colourMore Product Information