Wall - mounted safety cabinets, Sure-Grip® EX
Supplier: JUSTRITE MANUFACTURING
8917008
JUSU8917008EA 2510 SGD
JUSU8917008
Wall - mounted safety cabinets, Sure-Grip® EX
Furniture Storage Cabinets
Undercounter cabinet provides quick access at workstations by taking advantage of undercounter space for the safe storage of flammables. It comes with wall mounting hardware.
  • Eye-level positioning allows for easy access to solvents
Certifications: Compliance with NFPA and OSHA.