Safety cabinets, slimline, Sure-Grip® EX
Supplier: JUSTRITE MANUFACTURING
892223 892205 895422 895405 895423 895401 895403 895421
JUSU892223EA 2880 SGD
JUSU892223 JUSU892205 JUSU895422 JUSU895405 JUSU895423 JUSU895401 JUSU895403 JUSU895421
Safety cabinets, slimline, Sure-Grip® EX
Furniture Storage Cabinets
These slimline cabinets used alone or alongside larger cabinets for expanded storage when floor space is limited. Cabinet includes three galvanised steel shelves and a 51 mm leakproof sump for spill control. It includes dual vents, adjustable leveling feet, a grounding connector and large trilingual warning label.
  • Fire resistant
  • Stores flammable fuels and chemicals
  • Double-walled, air insulated and welded steel construction
Certifications: Meets OSHA and NFPA compliance with FM approval.
Order Now
SPECIFICATIONS
Environmentally Preferable