Sure-Grip® EX Safety Cabinets for Corrosives
Supplier: JUSTRITE MANUFACTURING
893022 894582
JUSU893022EA 1650 SGD
JUSU893022 JUSU894582
Sure-Grip® EX Safety Cabinets for Corrosives
Furniture Storage Cabinets
Steel cabinets store corrosive liquids and acids safely and securely, protecting both personnel and facilities.
  • U-Loc™ handles that include two keys and accept padlocks
  • Four adjustable levelling feet ensure a wobble-free base
  • Polyethylene trays are included to protect steel shelves from harsh chemicals
Exclusive Sure-Grip® EX features include Haz-Alert™ reflective exterior labelling for easy identification in fire conditions or power outages, and galvanized steel SpillSlope™ shelves that safely direct all spills to the 5,1 cm (2") leakproof sump.More Product Information