Safety disposal cans, quick-disconnect
Supplier: JUSTRITE MANUFACTURING
PP12755 TF12752 TF12755 PP12752 BY12752 BY12755 12160
JUSUPP12755EA 849 SGD
JUSUPP12755 JUSUTF12752 JUSUTF12755 JUSUPP12752 JUSUBY12752 JUSUBY12755 JUSU12160
Safety disposal cans, quick-disconnect
Safety Cans
Quick-disconnect safety disposal cans are the safe, compliant way to collect HPLC waste. Standardized configurations with a choice of stainless steel or polypropylene fittings or a combination of the two.
PE.
  • Hands-free solvent collection
  • Secure, easy connection and disconnection
  • Durable and safe with a maximum usage of manifolds enable upto 12 waste lines to be fed into a single intake fitting
Order Now
SPECIFICATIONS
Environmentally Preferable