Powder Funnels, PP
Supplier: KARTELL
0016600
KART0016600EA 51.7 SGD
KART0016600
Powder Funnels, PP
Funnels Powder Funnels
60° angle, autoclavable funnels.
  • Large bore avoids bridging of powders during transfer
  • Ribbed outside to avoid pressure lock
  • Excellent chemical resistance
Can be used with liquids.
Certifications: The raw materials used comply with food-contact EU regulation 1935/2004, 10/2011, US Food and Drug Administration and REACH/ROHS regulation.
Order Now
SPECIFICATIONS
Environmentally Preferable