Autoclave Sterilisation Tape
Supplier: KARTELL
1052
KART1052EA 22.4 SGD
KART1052
Autoclave Sterilisation Tape
Sterilisation Indicators
Adhesive tape.
  • Totally lead free
Specially designed for steam sterilisation (20 minutes at 120 °C, 10 minutes at 125 °C or 5 minutes at 135 °C). Colour change from green to dark green/black.
Shelf life: 36 months