Sedimentation cones, Imhoff type
Supplier: KARTELL
1055
KART1055EA 56.1 SGD
KART1055
Sedimentation cones, Imhoff type
Funnels Sedimentation Cones
Made of Acrylic, with PP screw cap.
  • Permanently moulded graduations
  • Clear
  • Easily drained via screw plug in the base