Pipette controller, Seropet
Supplier: KARTELL
1295.10
KART1295.10EA 2450 SGD
KART1295.10
Pipette controller, Seropet
Pipettes Pipette Controllers
Seropet was developed for aspirating liquids by glass or plastic pipettes ranging from the volume of 0,5 ml up to 100 ml.
  • Two different work modes: quick and normal, with continuous or pulsating adjustment
  • Particularly light structure ensures comfortable use even when used for a long time
  • Supplied with safety valve to block aspiration excesses
  • Equipped with autoclavable protective hydrophobic anti-aerosol filter
  • Highly sensitive internal valves to get precision also with small volumes
  • NiMH battery supplied has an autonomy of up to 8 hours with continuous
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable