Holders for test tubes
Supplier: KARTELL
130 131 135
KART130EA 11.8 SGD
KART130 KART131 KART135
Holders for test tubes
Racks Tube Racks
PP, white.
  • Good thermal and chemical resistance
  • Can be used in water baths
  • The tubes contents are always visible
Designed for titrations.