Pipette filler
Supplier: KARTELL
198
KART198EA 124 SGD
KART198
Pipette filler
Pipettes Pipette Controllers
Pipette filler with plunger, will handle pipettes up to 10 ml.
  • Comfortable grip
  • Simply press plunger and release to fill pipette
  • Knurled cap adjusts the plunger stroke for repeating pre-set volume
  • Rubber chuck will hold pipette securely and safely
  • Graduations on the body
  • Moulded in yellow ABS
Order Now
SPECIFICATIONS
Environmentally Preferable