Pipette bulb, with adapter
Supplier: KARTELL
200
KART200EA 22.1 SGD
KART200
Pipette bulb, with adapter
Pipettes Pipette Bulbs
Moulded pipette bulb made of synthetic rubber.
  • Three glass ball valves
  • Can be controlled by finger pressure
  • No metal parts to corrode
  • Can be used with all liquids except those that attack rubber
  • Use on bulb or cylindrical pipettes of different capacities
Delivery information: Supplied with adapter for pipettes with capacities more than 20 ml.
Order Now
SPECIFICATIONS
Environmentally Preferable