Magnetic Stirrer, explosion proof
Supplier: KARTELL
203
KART203EA 245 SGD
KART203
Magnetic Stirrer, explosion proof
Stirrers Magnetic Stirrers
Magnetic stirrer with ABS housing that has no electric motor but is powered by either air (1 to 2,5 kg/cm²; 15 to 40 psi) or water pressure.
  • Ideal for environments where flammable solvents and acid vapours are in high concentration
  • No electric motor so risk of sparking hazard is removed
  • Corrosion proof plastic construction
  • Can be used on the bench or clamper to a 12 mm Ø stand
Ordering information: Retort rods Ø 12 mm and stand to be ordered separately