Draining rack, wall mounted
Supplier: KARTELL
213
KART213EA 248 SGD
KART213
Draining rack, wall mounted
Racks Draining Racks
PS, gray.
  • The draining rods can be inserted and replaced as required
  • Supplied complete with drainage tube and fixtures for wall mounting
  • Optional rods for narrow-necked vessels are available to order under article number 212-6109
With integral extra wide draining channel and 72 draining rods which are 95 mm long, 15 mm Ø. The rod insertion apertures are closed at the back. This prevents water from seeping into the wall.