Eyewash station, First Eyd
Supplier: KARTELL
2384
KART2384EA 51.3 SGD
KART2384
Eyewash station, First Eyd
First Aid Supplies Eyewash Stations and Eyewash Bottles
Emergency eyewash station moulded in strong high impact PS. The bottle is either included or needs to be ordered seperately. It can be filled with distilled water or other medicated liquid for ophthalmic use.
  • Clearly printed instructions (in English or French) are printed on the support rack
  • Easy to attach to the wall
  • PE bottle with eye cup with snap on cap
  • Bottle is carefully designed to allow a quick and immediate flushing to remove foreign matters and prevent serious eye damage
Order Now
SPECIFICATIONS
Environmentally Preferable