Pipette stand
Supplier: KARTELL
255
KART255EA 33.4 SGD
KART255
Pipette stand
Pipette Stands
Stand made from PP, for 16 pipettes.
  • For 8 pipettes diameter <10 mm and 8 pipettes diameter <15 mm
  • Small holes in base allow drainage
W×D×H: 200×75×150 mm