Urine and biological specimen containers, with screw caps
Supplier: KARTELL
2596 2696 5630 5631
KART2596EA 398 SGD
KART2596 KART2696 KART5630 KART5631
Urine and biological specimen containers, with screw caps
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PS, transparent with PE or PP screw caps.
  • 60 ml with PE screw caps and 200 ml with PP screw caps
  • Quick opening and closing twist cap, leakproof
Ideal for collecting, storing and transporting urine samples and biological specimens.
* Aseptic containers individually wrapped, with label.
Order Now
SPECIFICATIONS
Environmentally Preferable