Pipette stand, rotary
Supplier: KARTELL
261
KART261EA 84 SGD
KART261
Pipette stand, rotary
Pipette Stands
Rotary pipette holder made from PP, for up to 94 pipettes.
  • Rotating base
  • Accommodates up to 94 pipettes with a diameter not exceeding 14 mm
Ø×H: 230×470 mm