Microtube racks
Supplier: KARTELL
296
KART296EA 21.6 SGD
KART296
Microtube racks
Racks Tube Racks
PP, white, for 20×1,5 ml reaction vessels.
  • Numbered positions to aid sample identification
  • Optional brackets can be used to join several racks together
  • Two levels for easy organisation