Centrifuge tubes
Supplier: KARTELL
2300 2302 300 301
KART2300EA 66 SGD
KART2300 KART2302 KART300 KART301
Centrifuge tubes
Tubes Centrifuge Tubes
PP, conical bottom, autoclavable.
  • Translucent, excellent chemical resistance
  • Can withstand temperatures up to +120 °C