Spray bottle
Supplier: KARTELL
327
KART327EA 53.1 SGD
KART327
Spray bottle
Bottles Spray Bottles
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • Suitable for foodstuff
Spray bottle atomiser with adjustment jet.