Dispensing bottle
Supplier: KARTELL
328
KART328EA 25.4 SGD
KART328
Dispensing bottle
Bottles Spray Bottles
HDPE/PP.
  • Good chemical resistance
  • Good temperature resistance
Dispensing bottle suitable for liquids single and/or continuous dispensing.
Order Now
SPECIFICATIONS
Environmentally Preferable