Staining racks
Supplier: KARTELL
354
KART354EA 17.4 SGD
KART354
Staining racks
Racks Staining Racks
PP.
  • For use with 631-9511
Staining rack with horizontal tray, holds 20 standard slides 76×26 mm.
W×D×H: 86×695×21 mm