Replacement spigot for carboys and aspirator bottles
Supplier: KARTELL
375
KART375EA 9.7 SGD
KART375
Replacement spigot for carboys and aspirator bottles
Bottles Carboys
HDPE, white.
  • Good chemical resistance
  • Leakproof
  • Suitable for carboys/aspirator bottles 216-5744 to 216-5747
With ¾" BSP screw thread.