Dropping bulbs for pipettes and tubes
Supplier: KARTELL
3914
KART3914EA 1.5 SGD
KART3914
Dropping bulbs for pipettes and tubes
Pipettes Pipette Bulbs
Dropping bulbs with rim, made of latex. Suitable for Ranvier dropping tubes and Pasteur pipettes.