Slide dispenser
Supplier: KARTELL
540
KART540EA 67.8 SGD
KART540
Slide dispenser
Slide Holders
This dispenser can accommodate up to 60 standard microscopy slides.
  • Fingerprint-free dispensing by rotating the knurled knobs on either side of the moulded base
  • Cover protecting the slides from dust
  • Rubber rollers to ease out the slide, then handled by the edges