Test tube racks
Supplier: KARTELL
370 559 560 562
KART370EA 14.3 SGD
KART370 KART559 KART560 KART562
Test tube racks
Racks Tube Racks
PP, white.
  • Excellent chemical resistance
  • Heat resistant up to 121 °C, autoclavable
  • Light, very robust
With two or three tiers.