Racks, microcentrifuge tube
Supplier: KARTELL
563-03 563-04 563-10 563-06
KART563-03EA 17.5 SGD
KART563-03 KART563-04 KART563-10 KART563-06
Racks, microcentrifuge tube
Racks Tube Racks
PP, autoclavable.
  • Versatile racks, can be used dry, in water baths or in freezers
  • Stackable when empty or full
  • Alphanumeric grid reference for easy sample identification
Can accommodate 100×1,5 ml microtubes with closed caps or without caps, or 50×1,5 ml microtubes with caps open.