Urine sample containers, with screw cap
Supplier: KARTELL
5640 5641 5571 5642 5643 5644
KART5640EA 175 SGD
KART5640 KART5641 KART5571 KART5642 KART5643 KART5644
Urine sample containers, with screw cap
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
PP, translucent urine cups with screw cap. Non sterile cups have a blue screw cap and are sold in multi-packs. Sterile cups have a red screw cap and a satinised marking area and are individually wrapped.
  • Available sterile or non sterile
  • Quick opening and closing twist cap, leakproof
Order Now
SPECIFICATIONS
Environmentally Preferable