Biohazard Transport Carriers
Supplier: KARTELL
569
KART569EA 237 SGD
KART569
Biohazard Transport Carriers
Carriers Test Tubes Transport Carriers
Transparent PC, silicone gasket and stainless steel handle.
  • Completely autoclavable
  • Biohazard symbol on the box
  • User sheet with instructions approved by WHO
To carry tubes or urine stool containers under safe conditions.