Magnetic stirring bars, PTFE coated, ball shaped
Supplier: KARTELL
770
KART770EA 12 SGD
KART770
Magnetic stirring bars, PTFE coated, ball shaped
Stirrers Magnetic Stirring Bars
Magnetic stirrers which are ideal for test tubes or for eccentric stirring movements.
  • Suitable for use with foodstuffs