Stirring rods
Supplier: KARTELL
828
KART828EA 18.3 SGD
KART828
Stirring rods
Stirrers Stirring Rods
White, one-piece rod with flattened ends which form two small spatulas.
  • Dimensions of spatulas (W×H) 15×12 and 12×9 mm
Dimensions of spatulas (W×H) 15×12 and 12×9 mm.