LDPE Bucket, 610/615
Supplier: Kautex
2000070588
KAUT2000070588EA 81.1 SGD
KAUT2000070588
LDPE Bucket, 610/615
Buckets
All buckets have strong walls and are very stable for a long service life.
  • Thick-walled, stable industrial and laboratory bucket
  • Stainless steel carrying handle covered with plastic
  • Interior graduations
  • Halal certified
Buckets feature a a liter scale on the inside. The laboratory buckets with spout have a steel carrying handle covered with plastic, and are suitable for food contact.