Screw caps for bottles with narrow neck, 301 and 302 series
Supplier: Kautex
2000090604
KAUT2000090604EA 0.6 SGD
KAUT2000090604
Screw caps for bottles with narrow neck, 301 and 302 series
Closures Bottle Closures
LDPE, translucent.
  • Good chemical and thermal resistance
  • Leakproof
For narrow neck bottles in the 301 and 302 series.