Containers, wide mouth, square with thread
Supplier: Kautex
310-70597 310-70598 310-70599 310-70699 310-70700 310-70701 310-73352 310-73353 310-70596 310-70698 310-70595 310-70697
KAUT310-70597EA 1.8 SGD
KAUT310-70597 KAUT310-70598 KAUT310-70599 KAUT310-70699 KAUT310-70700 KAUT310-70701 KAUT310-73352 KAUT310-73353 KAUT310-70596 KAUT310-70698 KAUT310-70595 KAUT310-70697
Containers, wide mouth, square with thread
Sample Containers General Purpose Containers
PVC, square, transparent or brown transparent colours.
  • Wide neck opening
  • Easy to fill, easy to clean
  • Leakproof
  • Suitable for food
  • Halal certified
Due to the wide neck opening the containers are easy to fill and to clean. Black closure with PE-foam insert. Brown transparent containers are suitable for light-sensitive media.
Order Now
SPECIFICATIONS
Environmentally Preferable