Containers, wide mouth, square, threaded
Supplier: Kautex
310-84412
KAUT310-84412EA 88.2 SGD
KAUT310-84412
Containers, wide mouth, square, threaded
Sample Containers General Purpose Containers
PVC. Square. Transparent or brown transparent colours.
  • Wide neck opening
  • Easy to fill and to clean
  • Leakproof
  • Suitable for food
  • With tamper-evident closure, PP, black colour
  • Halal certified
Brown transparent containers are suitable for light-sensitive media. Due to the wide neck opening the containers are easy to fill and to clean. The contents are safe against unauthorized removal. Opening the closure leaves distinct traces.