Buckets
Supplier: Kautex
610-70589
KAUT610-70589EA 96.6 SGD
KAUT610-70589
Buckets
Buckets
HDPE, with graduation.
  • Rustproof handle
  • Thick-walled
  • Sturdy
  • Suitable for food
  • Halal certified
Industrial and laboratory buckets are available with and without spout. Plastic coated steel handle. Builder’s buckets are dark grey with a rustproof handle. All buckets are thick-walled and very stable for a long working life and with graduated litre scale on the inside.