Weighing Bottles, Glass
Supplier: Kavalier
632421202804 632421202503
KAVA632421202804EA 188 SGD
KAVA632421202804 KAVA632421202503
Weighing Bottles, Glass
Bottles Weighing Bottles
Made of borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance