Dewar vessels, cylindrical, type C and G-C
Supplier: KGW
1011 10111 10112 10114 10115 10117 10118 1012 10120 10122 10123 10124 1013 1014 1015 1016 1018 1019 1021 10211 10212 10214 10215 10217 10218 1022 10220 10222 10223 10224 1023 1024 1025 1026 1028 1029 1061 10610 10611 10612 10613 10614 1063 1064 1066 1067 1068 10616 10615 10213 10216 10221 10210 1027 10113
KGWA1011EA 144 SGD
KGWA1011 KGWA10111 KGWA10112 KGWA10114 KGWA10115 KGWA10117 KGWA10118 KGWA1012 KGWA10120 KGWA10122 KGWA10123 KGWA10124 KGWA1013 KGWA1014 KGWA1015 KGWA1016 KGWA1018 KGWA1019 KGWA1021 KGWA10211 KGWA10212 KGWA10214 KGWA10215 KGWA10217 KGWA10218 KGWA1022 KGWA10220 KGWA10222 KGWA10223 KGWA10224 KGWA1023 KGWA1024 KGWA1025 KGWA1026 KGWA1028 KGWA1029 KGWA1061 KGWA10610 KGWA10611 KGWA10612 KGWA10613 KGWA10614 KGWA1063 KGWA1064 KGWA1066 KGWA1067 KGWA1068 KGWA10616 KGWA10615 KGWA10213 KGWA10216 KGWA10221 KGWA10210 KGWA1027 KGWA10113
Dewar vessels, cylindrical, type C and G-C
Dewars and Cryogenic Vessels
Dewar vessel, in silvered borosilicate glass, blue coated metal protective covering. For storage of liquid nitrogen, dry ice and other coolants.
Order Now
SPECIFICATIONS
Model 00 A 9 A 10 A 12 A 13 A 15 A 16 A 0 A 18 A 20 A 21 A 22 A 11 A 2 A 3 A 4 A 6 A 7 A 00 C 9 C 10 C 12 C 13 C 15 C 16 C 0 C 18 C 20 C S 21 C S 22 C 1 C 2 C 3 C 4 C 6 C 7 C G 0 C G 12 C G 13 C G 15 C G 16 C G 18 C G 2 C G 3 C G 6 C G 7 C G 9 C GS 21 C G 20 C 11 C 14 C 19 C 8 C 5 C 11 A
Capacity 0,1 L 1 L 1,5 L 1,5 L 2 L 1,5 L 2 L 0,2 L 2,5 L 3 L 4 L 8 L 0,3 L 0,45 L 0,5 L 0,75 L 0,8 L 1,2 L 0,1 L 1 L 1,5 L 1,5 L 2 L 1,5 L 2 L 0,2 L 2,5 L 3 L 4 L 8 L 0,3 L 0,45 L 0,5 L 0,75 L 0,8 L 1,2 L 0,2 L 1,5 L 2 L 1,5 L 2 L 2,5 L 0,45 L 0,5 L 0,8 L 1,2 L 1 L 4 L 3 L 2,1 L 3,2 L 5 L 1,7 L 1,25 L 2,1 L
Environmentally Preferable
Temperature range −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200 ...+150 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200...+200 °C −200 ...+150 °C
Daily evaporation 0,3 L 0,35 L 0,4 L 0,5 L 0,5 L 0,7 L 0,6 L 0,2 L 0,7 L 1,2 L 1,0 L 0,9 L 0,2 L 0,25 L 0,35 L 0,3 L 0,35 L 0,3 L 0,2 L 0,5 L 0,5 L 0,7 L 0,6 L 0,7 L 0,25 L 0,35 L 0,35 L 0,3 L 0,35 L 1,0 L 1,2 L 0,4 L 0,45 L 0,6 L 0,3 L 0,25 L
Int.Ø 40 mm 77 mm 77 mm 90 mm 90 mm 100 mm 100 mm 40 mm 110 mm 138 mm 138 mm 138 mm 47 mm 47 mm 57 mm 57 mm 67 mm 67 mm 40 mm 77 mm 77 mm 90 mm 90 mm 100 mm 100 mm 40 mm 110 mm 138 mm 138 mm 138 mm 47 mm 47 mm 57 mm 57 mm 67 mm 67 mm 40 mm 90 mm 90 mm 100 mm 100 mm 110 mm 47 mm 57 mm 67 mm 67 mm 77 mm 138 mm 138 mm 77 mm 90 mm 110 mm 67 mm 57 mm 77 mm
Int. height 90 mm 235 mm 345 mm 245 mm 340 mm 240 mm 290 mm 170 mm 290 mm 230 mm 310 mm 600 mm 190 mm 270 mm 210 mm 310 mm 240 mm 350 mm 90 mm 235 mm 345 mm 245 mm 340 mm 240 mm 290 mm 170 mm 290 mm 230 mm 310 mm 600 mm 190 mm 270 mm 210 mm 310 mm 240 mm 350 mm 170 mm 245 mm 340 mm 240 mm 290 mm 290 mm 270 mm 210 mm 240 mm 350 mm 235 mm 310 mm 230 mm 130 mm 600 mm 600 mm 500 mm 500 mm 500 mm
Overall height 135 mm 290 mm 395 mm 300 mm 390 mm 300 mm 345 mm 215 mm 345 mm 280 mm 375 mm 660 mm 240 mm 310 mm 260 mm 370 mm 295 mm 395 mm 215 mm 300 mm 390 mm 300 mm 345 mm 345 mm 310 mm 260 mm 295 mm 395 mm 290 mm 375 mm 280 mm 550 mm 665 mm 660 mm 560 mm 550 mm