Dish-shaped Dewar flasks, type SCH-CAL
Supplier: KGW
1191 1192 1193 1194 1195 1196 1197 1198 1671 1675 1674 1672
KGWA1191EA 347 SGD
KGWA1191 KGWA1192 KGWA1193 KGWA1194 KGWA1195 KGWA1196 KGWA1197 KGWA1198 KGWA1671 KGWA1675 KGWA1674 KGWA1672
Dish-shaped Dewar flasks, type SCH-CAL
Dewars and Cryogenic Vessels
Silvered borosilicate glass, structured aluminium protective covering. For tempering of round bottom flasks, cooling of small samples. They are vacuum insulated and silvered.
  • Suitable as hot or cold bath
  • Can be used on a magnetic stirrer
  • The cover is anti-magnetic and stainless
Temperature range from −200 to +200 °C.
CAL flasks have an external stucco aluminium protective casing, alternatively the glass inserts (without casing) are also available.