Dewar flasks, type GSS
Supplier: KGW
2300 2302 2303 2304 2305 2680 2681 2682 2301
KGWA2300EA 976 SGD
KGWA2300 KGWA2302 KGWA2303 KGWA2304 KGWA2305 KGWA2680 KGWA2681 KGWA2682 KGWA2301
Dewar flasks, type GSS
Dewars and Cryogenic Vessels
Dewar flasks in stainless steel for storage and transport of liquid nitrogen, dry ice and other coolants. They are vacuum insulated and have a temperature range from −200 to +200 °C.
  • With side grip and transport handle (except model GSS 500)